CAS RN 29840-57-1
IUPAC Name:
ethyl 5-aminopentanoate hydrochloride
Synonyme:
ethyl 5-aminopentanoate hydrochloride
Molekulargewicht (g/mol):
181.66
Summenformel:
C7H16ClNO2
InChi Key:
JELXJZASXLHKDV-UHFFFAOYSA-N
SMILES:
Cl.CCOC(=O)CCCCN
1
–
1
von
1
Ergebnisse
1
Ethyl5-Aminovalerat-Hydrochlorid, 96%, Thermo Scientific Chemicals
CAS: 29840-57-1 Summenformel: C7H16ClNO2 Molekulargewicht (g/mol): 181.66 MDL-Nummer: MFCD12546205 InChI-Schlüssel: JELXJZASXLHKDV-UHFFFAOYSA-N Synonym: ethyl 5-aminovalerate hydrochloride,ethyl 5-aminopentanoate hydrochloride,ethyl 5-aminovalerate hcl,5-aminovaleric acid ethyl ester hydrochloride,pentanoic acid, 5-amino-, ethyl ester, hydrochloride,ethyl 5-aminopentanoate hcl,ethyl5-aminovaleratehydrochloride,ethyl 5-azanylpentanoate hydrochloride,5-amino-valeric acid ethyl ester hydrochloride,5-aminopentanoic acid ethyl ester hydrochloride PubChem CID: 16094861 IUPAC-Name: Ethyl-5-aminopentanoat;hydrochlorid SMILES: CCOC(=O)CCCCN.Cl