CAS RN 33632-74-5
IUPAC Name:
3,4-dihydro-2H-1,5-benzodioxepine-2-carboxylic acid
Synonyme:
3,4-dihydro-2H-1,5-benzodioxepine-2-carboxylic acid
Molekulargewicht (g/mol):
194.19
Summenformel:
C10H10O4
InChi Key:
BYYFEADAQZCIGM-UHFFFAOYNA-N
SMILES:
OC(=O)C1CCOC2=CC=CC=C2O1
1
–
1
von
1
Ergebnisse
1
3,4-Dihydro-2H-1,5-Benzodioxepin-7-Carbonsäure, 98 %, Thermo Scientific Chemicals
CAS: 33632-74-5 Summenformel: C10H10O4 Molekulargewicht (g/mol): 194.186 MDL-Nummer: MFCD00276310 InChI-Schlüssel: BYYFEADAQZCIGM-UHFFFAOYSA-N Synonym: 3,4-dihydro-2h-benzo b 1,4 dioxepine-2-carboxylic acid,2h-1,5-benzodioxepin-2-carboxylicacid, 3,4-dihydro,3,4-dihydro-2h-1,5-benzodioxepine-2-carboxylic acid,2h-1,5-benzodioxepin-2-carboxylic acid, 3,4-dihydro,3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylicacid PubChem CID: 53708627 IUPAC-Name: 3,4-Dihydro-2H-1,5-Benzodioxepin-4-Carbonsäure SMILES: C1COC2=CC=CC=C2OC1C(=O)O