CAS RN 374-98-1
CAS RN 374-98-1
IUPAC Name:
1,1,1,2,2,3,3-heptafluoro-4-iodobutane
Synonyme:
1,1,1,2,2,3,3-heptafluoro-4-iodobutane
Molekulargewicht (g/mol):
309.95
Summenformel:
C4H2F7I
InChi Key:
AFHPVXVEXINDFS-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(F)(F)C(F)(F)CI
1
–
1
von
1
Ergebnisse
1
2,2,3,3,4,4,4-Heptafluor-1-Iodbutan, 96 %, Thermo Scientific Chemicals
CAS: 374-98-1 Summenformel: C4H2F7I Molekulargewicht (g/mol): 309.953 MDL-Nummer: MFCD00013719 InChI-Schlüssel: AFHPVXVEXINDFS-UHFFFAOYSA-N Synonym: heptafluorobutyl iodide,2,2,3,3,4,4,4-heptafluoro-1-iodobutane,butane, 1,1,1,2,2,3,3-heptafluoro-4-iodo,1h,1h-heptafluorobutyl iodide,1h,1h-heptafluoro-1-iodobutane,acmc-20aoko,1-iodo-1h,1h-perfluorobutane,1h,1h-heptafluoro-1-iodobutan,1-iodo-2,2,3,3,4,4,4-heptafluorobutane PubChem CID: 67806 IUPAC-Name: 1,1,1,2,2,3,3-Heptafluor-4-Iodbutan SMILES: C(C(C(C(F)(F)F)(F)F)(F)F)I