CAS RN 477251-98-2
IUPAC Name:
[5-(methylsulfanyl)pyridin-3-yl]boronic acid
Synonyme:
5-(methylsulfanyl)pyridin-3-ylboronic acid
Molekulargewicht (g/mol):
169.01
Summenformel:
C6H8BNO2S
InChi Key:
QNLQSEKMIZSQOG-UHFFFAOYSA-N
SMILES:
CSC1=CN=CC(=C1)B(O)O