CAS RN 497147-93-0
CAS RN 497147-93-0
IUPAC Name:
(5-cyanopyridin-3-yl)boronic acid
Synonyme:
5-cyanopyridin-3-ylboronic acid
Molekulargewicht (g/mol):
147.93
Summenformel:
C6H5BN2O2
InChi Key:
CYEXXDYQJPRMIQ-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(=CN=C1)C#N