CAS RN 506-13-8
IUPAC Name:
16-hydroxyhexadecanoic acid
Synonyme:
16-hydroxyhexadecanoic acid juniperic acid ω-hydroxypalmitic acid
Molekulargewicht (g/mol):
272.43
Summenformel:
C16H32O3
InChi Key:
UGAGPNKCDRTDHP-UHFFFAOYSA-N
SMILES:
OCCCCCCCCCCCCCCCC(O)=O
1
–
2
von
2
Ergebnisse
1
16-Hydroxyhexadecansäure, 98 %, Thermo Scientific Chemicals
CAS: 506-13-8 Summenformel: C16H32O3 Molekulargewicht (g/mol): 272.429 MDL-Nummer: MFCD00002750 InChI-Schlüssel: UGAGPNKCDRTDHP-UHFFFAOYSA-N Synonym: juniperic acid,16-hydroxypalmitic acid,hexadecanoic acid, 16-hydroxy,palmitic acid, 16-hydroxy,omega-hydroxypalmitic acid,16-hydroxy-hexadecanoic acid,unii-7ipp3u0f3i,.omega.-hydroxypalmitic acid,7ipp3u0f3i,16-hydroxy hexadecanoic acid PubChem CID: 10466 ChEBI: CHEBI:55328 IUPAC-Name: 16-Hydroxyhexadecansäure SMILES: C(CCCCCCCC(=O)O)CCCCCCCO