CAS RN 53064-79-2
IUPAC Name:
iodomethyl 2,2-dimethylpropanoate
Synonyme:
iodomethyl 2,2-dimethylpropanoate
Molekulargewicht (g/mol):
242.06
Summenformel:
C6H11IO2
InChi Key:
PELJISAVHGXLAL-UHFFFAOYSA-N
SMILES:
CC(C)(C)C(=O)OCI
1
–
1
von
1
Ergebnisse
1
Iodomethyl-Pivalat, TRC
CAS: 53064-79-2 Summenformel: C6 H11 I O2 Molekulargewicht (g/mol): 242.05 Synonym: 2,2-Dimethyl-propanoic Acid Iodomethyl Ester,(Pivaloyloxy)methyl Iodide,2,2-Dimethylpropionic Acid Iodomethyl Ester,Iodomethyl 2,2-Dimethylpropanoate IUPAC-Name: Iodomethyl-2,2-Dimethylpropanoat SMILES: CC(C)(C)C(=O)OCI