CAS RN 568580-02-9
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
R&D Systems™ YM 254890
A Tocris product, part of R&D Systems. Gα-protein inhibitor; potent inhibitor of Gq/11 signaling