CAS RN 57462-42-7
CAS RN 57462-42-7
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
3
von
3
Ergebnisse
1
MedChemExpress [Nle11]-Substance P
MedChemExpress [Nle11]-Substance P is a substance P analog that avoids methionine oxidation problems.
2
MedChemExpress [Nle11]-Substance P
MedChemExpress [Nle11]-Substance P is a substance P analog that avoids methionine oxidation problems.
3
MedChemExpress [Nle11]-Substance P
MedChemExpress [Nle11]-Substance P is a substance P analog that avoids methionine oxidation problems.