CAS RN 6018-94-6
CAS RN 6018-94-6
IUPAC Name:
nickel(2+) dihydrate oxalate
Synonyme:
nickel(2+) dihydrate oxalate
Molekulargewicht (g/mol):
182.74
Summenformel:
C2H4NiO6
InChi Key:
VNYOIRCILMCTHO-UHFFFAOYSA-L
SMILES:
O.O.[Ni++].[O-]C(=O)C([O-])=O
1
–
1
von
1
Ergebnisse
1
Nickel(II)-Oxalat-Dihydrat, Thermo Scientific Chemicals
CAS: 6018-94-6 Summenformel: C2H4NiO6 Molekulargewicht (g/mol): 182.741 MDL-Nummer: MFCD00167155 InChI-Schlüssel: VNYOIRCILMCTHO-UHFFFAOYSA-L Synonym: nickel oxalate,nickel ii oxalate dihydrate,nickel oxalate dihydrate,acmc-1b92x,nickel 2+ ; oxalate; dihydrate,nickel oxalate,,nickel 2+ dihydrate oxalate,nickel 2+ ion dihydrate oxalate PubChem CID: 516789 IUPAC-Name: Nickel(2+);oxalat;dihydrat SMILES: C(=O)(C(=O)[O-])[O-].O.O.[Ni+2]