CAS RN 62-56-6
CAS RN 62-56-6
Thioharnstoff, +99 %, zur Analyse, Thermo Scientific Chemicals
CAS: 62-56-6 Summenformel: CH4N2S Molekulargewicht (g/mol): 76.12 MDL-Nummer: MFCD00008067 InChI-Schlüssel: UMGDCJDMYOKAJW-UHFFFAOYSA-N Synonym: thiocarbamide,2-thiourea,pseudothiourea,isothiourea,sulourea,2-thiopseudourea,sulfourea,thiuronium,thiocarbonic acid diamide,urea, thio PubChem CID: 2723790 ChEBI: CHEBI:36946 IUPAC-Name: thiourea SMILES: NC(N)=S
Acetophenon, 99 %, reinst, Thermo Scientific Chemicals
CAS: 62-56-6 Summenformel: CH4N2S Molekulargewicht (g/mol): 76.12 MDL-Nummer: MFCD00008067 InChI-Schlüssel: UMGDCJDMYOKAJW-UHFFFAOYSA-N Synonym: thiocarbamide,2-thiourea,pseudothiourea,isothiourea,sulourea,2-thiopseudourea,sulfourea,thiuronium,thiocarbonic acid diamide,urea, thio PubChem CID: 2723790 ChEBI: CHEBI:36946 IUPAC-Name: thiourea SMILES: NC(N)=S
Thioharnstoff, 99 %, Thermo Scientific Chemicals
CAS: 62-56-6 Summenformel: CH4N2S Molekulargewicht (g/mol): 76.12 MDL-Nummer: MFCD00008067 InChI-Schlüssel: UMGDCJDMYOKAJW-UHFFFAOYSA-N Synonym: thiocarbamide,2-thiourea,pseudothiourea,isothiourea,sulourea,2-thiopseudourea,sulfourea,thiuronium,thiocarbonic acid diamide,urea, thio PubChem CID: 2723790 ChEBI: CHEBI:36946 IUPAC-Name: thiourea SMILES: NC(N)=S
Thioharnstoff, ≥ 99 %, ACS Reagenz, Thermo Scientific Chemicals
CAS: 62-56-6 Summenformel: CH4N2S Molekulargewicht (g/mol): 76.12 MDL-Nummer: MFCD00008067 InChI-Schlüssel: UMGDCJDMYOKAJW-UHFFFAOYSA-N Synonym: thiocarbamide,2-thiourea,pseudothiourea,isothiourea,sulourea,2-thiopseudourea,sulfourea,thiuronium,thiocarbonic acid diamide,urea, thio PubChem CID: 2723790 ChEBI: CHEBI:36946 IUPAC-Name: thiourea SMILES: NC(N)=S
Thioharnstoff, ACS, min. 99 %, Thermo Scientific Chemicals
CAS: 62-56-6 Summenformel: CH4N2S Molekulargewicht (g/mol): 76.12 MDL-Nummer: MFCD00008067 InChI-Schlüssel: UMGDCJDMYOKAJW-UHFFFAOYSA-N Synonym: thiocarbamide,2-thiourea,pseudothiourea,isothiourea,sulourea,2-thiopseudourea,sulfourea,thiuronium,thiocarbonic acid diamide,urea, thio PubChem CID: 2723790 ChEBI: CHEBI:36946 IUPAC-Name: thiourea SMILES: NC(N)=S
Thioharnstoff, ExtraPure, SLR, Fisher Chemical
CAS: 62-56-6 Summenformel: CH4N2S Molekulargewicht (g/mol): 76.12 MDL-Nummer: MFCD00008067 InChI-Schlüssel: UMGDCJDMYOKAJW-UHFFFAOYSA-N Synonym: thiocarbamide,2-thiourea,pseudothiourea,isothiourea,sulourea,2-thiopseudourea,sulfourea,thiuronium,thiocarbonic acid diamide,urea, thio PubChem CID: 2723790 ChEBI: CHEBI:36946 IUPAC-Name: thiourea SMILES: NC(N)=S