CAS RN 6224-63-1
CAS RN 6224-63-1
IUPAC Name:
tris(3-methylphenyl)phosphane
Synonyme:
tris(3-methylphenyl)phosphane
Molekulargewicht (g/mol):
304.37
Summenformel:
C21H21P
InChi Key:
LFNXCUNDYSYVJY-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC=C1)P(C1=CC(C)=CC=C1)C1=CC=CC(C)=C1
1
–
1
von
1
Ergebnisse
1
Tri(m-tolyl)phosphin, 98+ %, Thermo Scientific Chemicals
CAS: 6224-63-1 Summenformel: C21H21P Molekulargewicht (g/mol): 304.373 MDL-Nummer: MFCD00008532 InChI-Schlüssel: LFNXCUNDYSYVJY-UHFFFAOYSA-N Synonym: tri-m-tolylphosphine,tris 3-methylphenyl phosphine,tri m-tolyl phosphine,phosphine, tris 3-methylphenyl,tris m-tolyl phosphine,phosphine, tri-m-tolyl,tris 3-methylphenyl phosphane,tris 3-tolyl phosphine,tri-m-tolyl-phosphane,tri-m-tolylphosphane PubChem CID: 80362 IUPAC-Name: tris(3-methylphenyl)phosphan SMILES: CC1=CC(=CC=C1)P(C2=CC=CC(=C2)C)C3=CC=CC(=C3)C