CAS RN 742112-33-0
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
R&D Systems™ OSU 03012
A Tocris product, part of R&D Systems. PDPK1 (PDK1) inhibitor; inhibits Akt signaling