CAS RN 7429-90-5
CAS RN 7429-90-5
Aluminiumdraht, 1.0 mm (0.04 Zoll) Durchmesser, geglüht, ≥99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminiumfolie, 0.1 mm (0.004 Zoll) Dicke, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC-Name: Aluminium SMILES: [Al]
Aluminiumkugel, 6.35 mm (0.25 Zoll) im Durchmesser x 6.35 mm (0.25 Zoll) Länge, Puratronic™, 99.9999 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC-Name: Aluminium SMILES: [Al]
Aluminiumkugel, 6.35 mm (0.25 Zoll) im Durchmesser x 12.7 mm (0.50 Zoll) Länge, Puratronic™, 99.999 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC-Name: Aluminium SMILES: [Al]
Aluminiumfloke, 11 Mikrometer, 99.7 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminiumgranulat, 8–12 mm (0.32–0.47 in.), 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminiumkugel, 6.35 mm (0.25 Zoll) im Durchmesser x 6.35 mm (0.25 Zoll) Länge, Puratronic™, 99.999 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC-Name: Aluminium SMILES: [Al]
Aluminiumfolie, 0.25 mm (0.01 Zoll) Dicke, geglüht, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminium-Kugel, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Dünne Aluminiumfolie, 2.0 Mikrometer (0.00008 Zoll) Dicke, 99.1 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminiumkugel, unregelmäßig, 15 mm und kleiner, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC-Name: Aluminium SMILES: [Al]
Aluminiumdraht, 99.99 % (Metallbasis), Thermo Scientific Chemicals, Puratronic™
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminiumfolie, 0.5 mm (0.02 Zoll) Dicke, geglüht, Puratronic™, 99.9995 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7429-90-5 Summenformel: Al Molekulargewicht (g/mol): 26.98 MDL-Nummer: MFCD00134029 InChI-Schlüssel: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC-Name: Aluminium SMILES: [Al]