CAS RN 7440-05-3
CAS RN 7440-05-3
Palladium auf Aktivkohle, 10 % Pd, nicht reduziert, trocken, Thermo Scientific Chemicals
CAS: 7440-05-3 Summenformel: Pd Molekulargewicht (g/mol): 106.42 MDL-Nummer: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI-Schlüssel: KDLHZDBZIXYQEI-UHFFFAOYSA-N Synonym: black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC-Name: Palladium SMILES: [Pd]
Palladium auf Aktivkohle, 10 % Pd, (50 % nass mit Wasser zur Sicherheit), unreduziert, Thermo Scientific Chemicals
CAS: 7440-05-3 Summenformel: Pd Molekulargewicht (g/mol): 106.42 MDL-Nummer: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI-Schlüssel: KDLHZDBZIXYQEI-UHFFFAOYSA-N Synonym: black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC-Name: Palladium SMILES: [Pd]
Palladium auf Aktivkohle, nicht reduziert, 5 % Pd, Thermo Scientific Chemicals
CAS: 7440-05-3 Summenformel: Pd Molekulargewicht (g/mol): 106.42 MDL-Nummer: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI-Schlüssel: KDLHZDBZIXYQEI-UHFFFAOYSA-N Synonym: black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC-Name: Palladium SMILES: [Pd]
Palladium auf Calciumcarbonat, vergiftet mit 3.5 % Blei, 5 % Pd, Thermo Scientific Chemicals
CAS: 7440-05-3 Summenformel: Pd Molekulargewicht (g/mol): 106.42 MDL-Nummer: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI-Schlüssel: KDLHZDBZIXYQEI-UHFFFAOYSA-N Synonym: black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC-Name: Palladium SMILES: [Pd]
Palladium-Standardlösung, für AAS, 1 mg/ml Pd in 10 % HCl, Thermo Scientific Chemicals
CAS: 7440-05-3 | Pd | 106.42 g/mol
Palladiumstandard, 1000 mg/l in 10 % HCl (Qualität: Assurance Grade), SPEX CertiPrep™
CAS: 7440-05-3 Summenformel: Pd Molekulargewicht (g/mol): 106.42 MDL-Nummer: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI-Schlüssel: KDLHZDBZIXYQEI-UHFFFAOYSA-N PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC-Name: Palladium SMILES: [Pd]
Palladium (5% on Alumina Powder, Reduced, Dry), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Palladium (5% on Alumina Powder, Reduced, Dry), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Palladium (5% on Barium Sulfate), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Palladium (5% on Calcium Carbonate, Lead poisoned), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Palladium (5% on Barium Sulfate), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Palladium On Carbon (extent of labeling: 10 wt. % loading, matrix carbon, dry support), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Palladium On Carbon (extent of labeling: 10 wt. % loading, matrix carbon, dry support), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Palladium (5% on Calcium Carbonate, Lead poisoned), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Palladium (5% on Barium Sulfate), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
- 1
- 2