CAS RN 7440-18-8
Ruthenium, 2 % auf 3.18 mm (0.125 Zoll)-Aluminiumoxidpellets, Thermo Scientific Chemicals
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Ruthenium, typischerweise 5 % auf Aluminiumoxidpulver, reduziert, Thermo Scientific Chemicals
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Ruthenium, 99.9 %, (Spurenmetallbasis), -200 Mesh, Pulver, Thermo Scientific Chemicals
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Rutheniumpulver, -22 Mesh, Premion , 99,98 % (Metallbasis, ohne Ca)
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Rutheniumpulver, 99. >99 % (Metallbasis ohne Ca), Thermo Scientific Chemicals, Premion™
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Rutheniumlösung für ICP/MS 10 mg/l HCl-Matrix, Fisher Chemical™
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Ruthenium-Standardlösung, für AAS, 1 mg/ml Ru in 5 % HCl, Thermo Scientific Chemicals
CAS: 7440-18-8 | Molecular Formula: Ru | Molecular Weight: 101.07 g/mol
Rutheniumpulver, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Ruthenium, 4 % auf 6.35 mm (0.25 Zoll)-Aluminiumoxidringen, Thermo Scientific Chemicals
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Ruthenium schwarz, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Ruthenium, 0.5 % auf granulärem Kohlenstoff, Thermo Scientific Chemicals
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Ruthenium, 5 % auf Aktivkohlepulver, reduziert, nominell 50 % Wasser nass, Thermo Scientific Chemicals
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]
Ruthenium für AAS 1 g/l HCl-Matrix, Fisher Chemical™
CAS: 7440-18-8 Summenformel: Ru Molekulargewicht (g/mol): 101.07 MDL-Nummer: MFCD00011207 MFCD03458417 InChI-Schlüssel: KJTLSVCANCCWHF-UHFFFAOYSA-N PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC-Name: Ruthenium SMILES: [Ru]