CAS RN 76461-17-1
CAS RN 76461-17-1
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
MedChemExpress Somatostatin-25
MedChemExpress Somatostatin-25 is a endogenous neuropeptide hormone that shows inhibitory activity against secretion of growth hormone.
2
MedChemExpress Somatostatin-25
MedChemExpress Somatostatin-25 is a endogenous neuropeptide hormone that shows inhibitory activity against secretion of growth hormone.