CAS RN 7783-71-3
IUPAC Name:
tantalum(5+) pentafluoride
Synonyme:
tantalum pentafluoride
Molekulargewicht (g/mol):
275.94
Summenformel:
F5Ta
InChi Key:
YRGLXIVYESZPLQ-UHFFFAOYSA-I
SMILES:
[F-].[F-].[F-].[F-].[F-].[Ta+5]
1
–
1
von
1
Ergebnisse
1
Tantal(V)-fluorid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7783-71-3 Summenformel: F5Ta Molekulargewicht (g/mol): 275.94 MDL-Nummer: MFCD00042542 InChI-Schlüssel: YRGLXIVYESZPLQ-UHFFFAOYSA-I Synonym: tantalum pentafluoride,tantalum v fluoride,tantalum fluoride,tantalum fluoride taf5,taf5 PubChem CID: 82218 IUPAC-Name: Pentafluortantal SMILES: F[Ta](F)(F)(F)F