CAS RN 902837-56-3
IUPAC Name:
ethyl 2-(2-chloropyridin-3-yl)-2-oxoacetate
Synonyme:
ethyl 2-(2-chloropyridin-3-yl)-2-oxoacetate
Molekulargewicht (g/mol):
213.62
Summenformel:
C9H8ClNO3
InChi Key:
ZTIMVJIGNBYGJG-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(=O)C1=C(Cl)N=CC=C1
1
–
1
von
1
Ergebnisse
1
Ethyl 2-Chlor-3-Pyridylglyoxylat, 97 %, Thermo Scientific Chemicals
CAS: 902837-56-3 Summenformel: C9H8ClNO3 Molekulargewicht (g/mol): 213.617 MDL-Nummer: MFCD07781194 InChI-Schlüssel: ZTIMVJIGNBYGJG-UHFFFAOYSA-N Synonym: ethyl 2-chloro-3-pyridylglyoxylate,ethyl 2-2-chloropyridin-3-yl-2-oxoacetate,ethyl 2-chloropyridin-3-yl oxo acetate,ethyl 2-chloro-3-pyridinyl oxoacetate,2-chloro-3-pyridylglyoxylic acid ethyl ester,ethyl 2-2-chloro 3-pyridyl-2-oxoacetate,2-chloro-pyridin-3-yl-oxo-acetic acid ethyl ester PubChem CID: 42553166 IUPAC-Name: Ethyl 2-(2-Chloropyridin-3-yl)-2-Oxoacetat SMILES: CCOC(=O)C(=O)C1=C(N=CC=C1)Cl