CAS RN 916734-43-5
CAS RN 916734-43-5
IUPAC Name:
2-{[3-(benzenesulfonyl)-5,5-dimethyl-1,3-thiazolidin-4-yl]formamido}-3-[(benzylcarbamoyl)amino]propanoic acid
Synonyme:
2-{[3-(benzenesulfonyl)-5,5-dimethyl-1,3-thiazolidin-4-yl]formamido}-3-[(benzylcarbamoyl)amino]propanoic acid
Molekulargewicht (g/mol):
520.62
Summenformel:
C23H28N4O6S2
InChi Key:
XKLHCUGVLCGKKX-UHFFFAOYNA-N
SMILES:
CC1(C)SCN(C1C(=O)NC(CNC(=O)NCC1=CC=CC=C1)C(O)=O)S(=O)(=O)C1=CC=CC=C1