New Feature
You can now browse the site in English or Italiano.
Controlled Environment Cleaners and Disinfectants product banner
Overhead Stirrers
Overhead stirrers are laboratory devices used to mix large-volume, viscous solutions. They typically include a stirrer, paddle, and stand. The Fisher Scientific channel offers a wide range of overhead stirrers with features such as digital displays, varied speed ranges, and advanced safety and control options.
IKA™ Eurostar Digital Laboratory Stirrer
Automatically adjusts the stirring speed through microprocessor controlled technology within the speed range of 0/30 - 2000 rpm.
IKA™ PT 100.53 1.4571 Stainless Steel Temperature Sensor
Velp Scientifica™ ES Overhead Stirrer
Analogical overhead stirrer with 15 Ncm torque, able to stir up to 15 l.
Velp Scientifica™ ES Overhead Stirrer
Heidolph™ Hei-TORQUE Core
The Hei-TORQUE Core is a light overhead stirrer with a torque of 40 Ncm.
Heidolph™ Hei-TORQUE Core
Selector Guide
Get the best-fit overhead stirrer for your lab protocols with our interactive selection guide.
Complete Your Order - Great Deals
Magnetic Stirrers
Magnetic stirrers are lab tools that use a rotating magnetic field to spin a stir bar and mix liquids. They support applications such as solution mixing, cell culture, and chemical reactions.
Magnetic Stirrers
Mechanical Homogenizers
Powered tools for grinding, lysing, and homogenizing samples before molecular extraction. Offered as handheld or benchtop units, with complete kits including motor, probe, and pestle accessories
Mechanical Homogenizers
General Purpose Wipers
General purpose wipers handle a range of cleaning, disinfection, and absorption tasks. Made from various materials in disposable or reusable formats, they come as individual wipes, bulk packs, or rolls.
General Purpose Wipers
FAQ
An overhead stirrer is a laboratory instrument that mixes liquids using a motor-driven shaft and impeller inserted directly into the sample.
Unlike magnetic stirrers, which rely on a magnetic field and are limited to low-viscosity fluids, overhead stirrers handle high-viscosity or large-volume mixtures up to several liters.
They are ideal for polymer synthesis, emulsification, suspensions, viscous reagents, and pilot-scale formulations where precise torque and continuous mixing are required.
  • Torque: Determines the stirrer’s ability to handle resistance; higher torque motors can process viscous or dense materials without speed loss.
  • Speed range: Variable-speed drives (typically 50–2 000 rpm) allow optimization for low-shear blending or high-shear dispersion.
  • Impeller type:
    • Propeller blades provide axial flow for homogeneous liquid-liquid mixing.
    • Anchor or paddle impellers suit high-viscosity pastes and gels.
    • Turbine or dispersing blades create strong radial shear for emulsions.
Selecting the correct impeller geometry and torque rating ensures efficient energy transfer, consistent shear, and reproducible results across batch sizes.
Modern overhead stirrers include advanced controls and safeguards for precision and laboratory safety:
  • Overload and stall protection to prevent motor damage under excessive load.
  • Digital torque feedback and closed-loop speed control to maintain constant mixing speed regardless of viscosity changes.
  • Soft start/stop and anti-splash acceleration to minimize sample loss.
  • Sealed housings (IP-rated) for protection against spills or corrosive vapors.
  • GLP/GMP-compliant logging, USB or RS-232 connectivity, and programmable profiles for traceability in regulated environments.
These features enhance reliability, user safety, and reproducibility for demanding analytical and process workflows.
Top Brands