missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17alpha-Ethynylestradiol-16,16-d2 3-Methyl Ether, CDN
Click to view available options
Menge:
0.005 g
0.01 g
Packungsgröße:
0.01 Gramm
Specifications
Specifications
| Chemischer Name oder Material | Mestranol-16,16-D2 |
| Summenformel | C21 D2 H24 O2 |
| Menge | 0.01 g |
| Synonym | 17 alpha-Ethynylestradiol-16,16-d2 3-Methyl Ether, (17α)-3-Methoxy-19-norpregna-1,3,5(10)-trien-20-yn-17-ol-16,16-d2, Mestranol-d2, Mestranol-16,16-D2, 17α-Ethinylestradiol-16,16-d2 3-methyl ether |
| InChI-Formel | InChI=1 S/C21H26O2/c1-4-21(22)12-10-19-18-7-5-14-13-15(23-3)6-8-16(14)17(18)9-11-20(19,21)2/h1,6,8,13,17-19,22 H,5,7,9-12H2,2-3H3/t17-,18-,19+,20+,21+/m1/s1/i12D2 |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CCc4cc(OC)ccc4[C@H]3 CC[C@]2(C)[C@]1(O)C#C |
| IUPAC-Name | (8 R,9 S,13 S,14 S,17 R)-16,16-dideuterio-17-ethynyl-3-methoxy-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-17-ol |
| Molekulargewicht (g/mol) | 312.4423 |
| Formelmasse | 312.2058 g/mol |
| Reinheit (%) | 97 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction