Learn More
5'-(Methyl-d3-thio)adenosine, CDN
Artikelnummer. 30316513
Click to view available options
Menge:
0.005 g
0.01 g
Unit Size:
0.01 Gramm
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316513
Brand: CDN CDND80020.01G
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemischer Name oder Material 5'-(Methylthio)adenosine-D3 (S-methyl-D3)
CAS 2699607-01-5
Summenformel C11 D3 H12 N5 O3 S
Menge 0.01 g
Synonym 5'-S-(Methyl-d3)-5'-thioadenosine, 5'-(Methyl-d3-thio)-5'-deoxyadenosine, 5'-(Methylthio)adenosine-D3 (S-methyl-D3), 5'-Deoxy(methylthio)adenosine-d3, 5'-Deoxy-5'-(methylthio-d3)adenosine, 1-(6-Amino-9 H-purin-9-yl)-1-deoxy-5-S-(methyl-d3)-5-thio-β-D-ribofuranose
InChI-Formel InChI=1 S/C11H15N5O3S/c1-20-2-5-7(17)8(18)11(19-5)16-4-15-6-9(12)13-3-14-10(6)16/h3-5,7-8,11,17-18 H,2H2,1H3,(H2,12,13,14)/t5-,7-,8-,11-/m1/s1/i1D3
SMILES [2 H]C([2 H])([2 H])SC[C@H]1 O[C@H]([C@H](O)[C@@H]1 O)n2cnc3c(N)ncnc23
IUPAC-Name (2 R,3 R,4 S,5 S)-2-(6-aminopurin-9-yl)-5-(trideuteriomethylsulfanylmethyl)oxolane-3,4-diol
Molekulargewicht (g/mol) 300.352
Formelmasse 300.1084 g/mol
Reinheit (%) 99 atom % D, min 98% Chemical Purity