missing translation for 'onlineSavingsMsg'
Learn More

Folpet-d4, CDN

Artikelnummer. 30325944
Click to view available options
Menge:
0.005 g
0.01 g
Packungsgröße:
0.01 Gramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Für den Kauf dieses Produkts gelten möglicherweise Einschränkungen. Etwaige zusätzliche Unterlagen werden bei Bedarf von Ihnen angefordert.

Artikelnummer. 30325944

Marke: CDN CDND74860.005G

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Folpet D4
CAS 1327204-12-5
Summenformel C9 2H4 Cl3 N O2 S
Menge 0.005 g
Synonym Folpet D4
InChI-Formel InChI=1 S/C9H4Cl3NO2S/c10-9(11,12)16-13-7(14)5-3-1-2-4-6(5)8(13)15/h1-4 H/i1D,2 D,3 D,4 D
SMILES [2 H]c1c([2 H])c([2 H])c2C(=O)N(SC(Cl)(Cl)Cl)C(=O)c2c1[2 H]
IUPAC-Name 4,5,6,7-tetradeuterio-2-(trichloromethylsulfanyl)isoindole-1,3-dione
Molekulargewicht (g/mol) 300.58
Formelmasse 298.9279 g/mol
Reinheit (%) 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.