missing translation for 'onlineSavingsMsg'
Learn More

Methyl Parathion-d6 (dimethyl-d6), CDN

Artikelnummer. 30312066
Click to view available options
Menge:
0.01 g
0.05 g
Unit Size:
0.01 Gramm
0.05 Gramm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30312066

Brand: CDN CDND42890.01G

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Parathion-methylD6(dimethylD6)
CAS 96740-32-8
Summenformel C8 2H6 H4 N O5 P S
Menge 0.01 g
Synonym Parathion-methyl D6 (dimethyl D6), Phosphorothioic acid, O,O-di(methyl-d3) O-(4-nitrophenyl) ester (9 CI)
InChI-Formel InChI=1 S/C8H10NO5PS/c1-12-15(16,13-2)14-8-5-3-7(4-6-8)9(10)11/h3-6 H,1-2H3/i1D3,2D3
SMILES [2 H]C([2 H])([2 H])OP(=S)(Oc1ccc(cc1)[N+](=O)[O-])OC([2 H])([2 H])[2 H]
IUPAC-Name (4-nitrophenoxy)-sulfanylidene-bis(trideuteriomethoxy)-λ^{5}-phosphane
Molekulargewicht (g/mol) 269.24
Formelmasse 269.0394 g/mol
Reinheit (%) 99 atom % D, min 95% Chemical Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.