missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-3-(2-Methoxy-d3-phenoxy)-1,2-propanediol, CDN
missing translation for 'orderingAttributeHoverText'
Menge:
0.05 g
0.1 g
missing translation for 'unitSize'
0.05 Gramm
0.10 Gramm
Specifications
Specifications
| Chemischer Name oder Material | rac Guaifenesin-d3 |
| CAS | 1189924-85-3 |
| Summenformel | C10 2H3 H11 O4 |
| Menge | 0.05 g |
| InChI-Formel | InChI=1 S/C10H14O4/c1-13-9-4-2-3-5-10(9)14-7-8(12)6-11/h2-5,8,11-12 H,6-7H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1ccccc1OCC(O)CO |
| IUPAC-Name | 3-[2-(trideuteriomethoxy)phenoxy]propane-1,2-diol |
| Molekulargewicht (g/mol) | 201.23 |
| Formelmasse | 201.108 g/mol |
| Reinheit (%) | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction