missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',5,5'-Tetrabromobisphenol A-d10, (OH)2, CDN
Spezifikation
Spezifikation
| Chemischer Name oder Material | 3,3',5,5'-Tetrabromobisphenol A-d10 |
| CAS | 2732834-96-5 |
| Summenformel | C15 D10 H2 Br4 O2 |
| Menge | 0.05 g |
| Synonym | Tetrabromobisphenol A D10 (dimethyl D3, bisphenol-3,5-D2);Tetrabromobisphenol A D10, 4,4'-(1,1,1,3,3,3-Hexadeuterio-2,2-propanediyl)bis(2,6-dibromo-3,5-dideuteriophenol), 2,2',6,6'-Tetrabromobisphenol A (ring,chain-D10), 3,3',5,5'-Tetrabromobisphenol A (ring,dimethyl-D10), 3,3',5,5'-Tetrabromobisphenol A D10 (ring D4 dimethyl D6) |
| InChI-Formel | InChI=1 S/C15H12Br4O2/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8/h3-6,20-21 H,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c(Br)c(O)c(Br)c([2 H])c1C(c2c([2 H])c(Br)c(O)c(Br)c2[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC-Name | 2,6-dibromo-3,5-dideuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(3,5-dibromo-2,6-dideuterio-4-hydroxyphenyl)propan-2-yl]phenol |
| Molekulargewicht (g/mol) | 553.9322 |
| Formelmasse | 549.8198 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?