missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl-1,1,1',1'-d4-stilbestrol-3,3',5,5'-d4, CDN
Click to view available options
Menge:
0.01 g
0.05 g
Packungsgröße:
0.01 Gramm
0.05 Gramm
Specifications
Specifications
| Chemischer Name oder Material | trans-Diethyl-1,1,1',1'-stilbestrol-3,3',5,5'-d8 |
| CAS | 91318-10-4 |
| Summenformel | C18 2H8 H12 O2 |
| Menge | 0.05 g |
| Synonym | Phen-2,6-d2-ol, 4,4'-[(1 E)-1,2-di(ethyl-1,1-d2)-1,2-ethenediyl]bis-, Phen-2,6-d2-ol, 4,4'-[1,2-di(ethyl-1,1-d2)-1,2-ethenediyl]bis-, (E)- (9 CI), 4,4'-[(1 E)-1,2-Di(ethyl-1,1-d2)-1,2-ethenediyl]bis[phen-2,6-d2-ol], trans-Diethyl-1,1,1',1'-stilbestrol-3,3',5,5'-d8, (E)-Diethyl-1,1,1',1'-d4-stilbestrol-3,3',5,5'-d4, Diethylstilbestrol (ring-3,3',5,5'-diethyl-1,1,1',1'-D8), Diethylstilbestrol (ring-3,3',5,5'-diethyl-1,1,1',1'-D8), Diethylstilbestrol (ring-3,3',5,5'-diethyl-1,1,1',1'-D8) |
| InChI-Formel | InChI=1 S/C18H20O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,19-20 H,3-4H2,1-2H3/b18-17+/i3D2,4D2,9 D,10 D,11 D,12 D |
| SMILES | [2 H]c1cc(cc([2 H])c1O)\C(=C(\c2cc([2 H])c(O)c([2 H])c2)/C([2 H])([2 H])C)\C([2 H])([2 H])C |
| IUPAC-Name | 2,6-dideuterio-4-[(E)-2,2,5,5-tetradeuterio-4-(3,5-dideuterio-4-hydroxyphenyl)hex-3-en-3-yl]phenol |
| Molekulargewicht (g/mol) | 276.4 |
| Formelmasse | 276.1965 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction