missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Nonyl-6,6,7,7-d4 Alcohol, CDN
Click to view available options
Menge:
0.05 g
0.1 g
Packungsgröße:
0.05 Gramm
0.10 Gramm
Spezifikation
Spezifikation
| Chemischer Name oder Material | 1-Nonanol-6,6,7,7-D4 |
| CAS | 33975-46-1 |
| Summenformel | C9 D4 H16 O |
| Menge | 0.05 g |
| Synonym | Nonyl-6,6,7,7-d4 alcohol (8 CI), 1-Nonanol-6,6,7,7-D4, Nonanol-6,6,7,7-d4, n-Nonyl-6,6,7,7-d4 alcohol |
| InChI-Formel | InChI=1 S/C9H20O/c1-2-3-4-5-6-7-8-9-10/h10H,2-9H2,1H3/i3D2,4D2 |
| SMILES | [2 H]C([2 H])(CC)C([2 H])([2 H])CCCCCO |
| IUPAC-Name | 6,6,7,7-tetradeuteriononan-1-ol |
| Molekulargewicht (g/mol) | 148.279 |
| Formelmasse | 148.177 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction