missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-Phenylephrine-2,4,6-d3 HCl, CDN
Click to view available options
Menge:
0.01 g
0.05 g
Unit Size:
0.01 Gramm
0.05 Gramm
Specifications
Specifications
| Chemischer Name oder Material | Phenylephrine-D3 (phenol-2,4,6-D3) Hydrochloride |
| CAS | 1276197-50-2 |
| Summenformel | C9 D3 H10 N O2 . Cl H |
| Menge | 0.05 g |
| Synonym | 3-[(1 R)-1-Hydroxy-2-(methylamino)ethyl]phenol-2,4,6-d3 hydrochloride, (αR)-3-Hydroxy-α-[(methylamino)methyl]benzene-2,4,6-d3-methanol hydrochloride, (1 R)-1-(3-Hydroxyphenyl-2,4,6-d3)-1-hydroxy-2-(methylamino)ethane hydrochloride, (-)-Phenylephrine-d3 hydrochloride, (1 R)-1-(3-Hydroxyphenyl-2,4,6-d3)-2-(N-methylamino)ethanol hydrochloride, N-[(2 R)-2-Hydroxy-2-(3-hydroxyphenyl-2,4,6-d3)ethyl]-N-methylamine hydrochloride, N-[(2 R)-2-Hydroxy-2-(3-hydroxyphenyl-2,4,6-d3)ethyl]-N-methylamine hydrochloride, N-[(2 R)-2-Hydroxy-2-(3-hydroxyphenyl-2,4,6-d3)ethyl]-N-methylamine hydrochloride, N-[(2 R)-2-Hydroxy-2-(3-hydroxyphenyl-2,4,6-d3)ethyl]-N-methylamine hydrochloride, N-[(2 R)-2-Hydroxy-2-(3-hydroxyphenyl-2,4,6-d3)ethyl]-N-methylamine hydrochloride |
| InChI-Formel | InChI=1 S/C9H13NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,9-12 H,6H2,1H3;1 H/t9-;/m0./s1/i3D,4 D,5 D; |
| SMILES | Cl.[2 H]c1cc([2 H])c([C@@H](O)CNC)c([2 H])c1O |
| IUPAC-Name | 2,4,6-trideuterio-3-[(1 R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride |
| Molekulargewicht (g/mol) | 206.6844 |
| Formelmasse | 206.0901 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction