Learn More
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-99138PE
This item is not returnable.
View return policy
Description
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Polyclonal antibody specifically detects Dimethylarginine Dimethylaminohydrolase 1/DDAH1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)