missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRE1 alpha, p Ser724 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB100-2323APC
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IRE1 alpha Polyclonal antibody specifically detects IRE1 alpha in Human, Mouse, Rat, Porcine, Drosophila, Goat, Mammalia, Monkey, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, In vitro Assay, ChIP assay (ChIP), KnockDown
Spezifikation
| IRE1 alpha | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, In vitro assay, Chromatin Immunoprecipitation (ChIP), Knockdown Validated | |
| EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, IRE1a, Ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1 | |
| This IRE1 alpha [p Ser724] antibody was raised against a synthetic peptide surrounding the phosphorylated serine 724 of the human IRE1 alpha protein. [Swiss-Prot #O75460] | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Diabetes Research, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuroscience, Signal Transduction, Unfolded Protein Response | |
| 2081 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, In vitro Assay, ChIP Assay, KnockDown | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Drosophila | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur