Learn More
Pyrrolidine-2,2,3,3,4,4,5,5-d8, CDN
missing translation for 'orderingAttributeHoverText'
Menge:
0.25 g
0.5 g
missing translation for 'unitSize'
0.25 Gramm
0.50 Gramm
Specifications
Specifications
| Chemischer Name oder Material | Pyrrolidine-d8 |
| CAS | 212625-79-1 |
| Summenformel | C4 D8 H N |
| Menge | 0.25 g |
| Synonym | Azacyclopentane-d8, Azolidine-d8, Butylenimine-d8, NSC 62781-d8, Perhydropyrrole-d8, Pyrrolidine-2,2,3,3,4,4,5,5-d8 (9 CI, ACI), Pyrrolidine-2,2,3,3,4,4,5,5-d8 (9 CI, ACI), Pyrrolidine-2,2,3,3,4,4,5,5-d8 (9 CI, ACI), Pyrrolidine-2,2,3,3,4,4,5,5-d8 (9 CI, ACI), Pyrrolidine-2,2,3,3,4,4,5,5-d8 (9 CI, ACI), Pyrrolidine-2,2,3,3,4,4,5,5-d8 (9 CI, ACI), Pyrrolidine-2,2,3,3,4,4,5,5-d8 (9 CI, ACI) |
| InChI-Formel | InChI=1 S/C4H9N/c1-2-4-5-3-1/h5H,1-4H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC-Name | 2,2,3,3,4,4,5,5-octadeuteriopyrrolidine |
| Molekulargewicht (g/mol) | 79.17 |
| Formelmasse | 79.1237 g/mol |
| Show More |