missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Ethylpiperidin-3-yl)methanamin, TRC

Codice prodotto. 30335506
Cambia vista
Click to view available options
Menge:
50 mg
Dimensione della confezione:
50 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Menge Dimensione della confezione
30335506 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30335506 Fornitore Toronto Research Chemicals N. del fornitore TRCB43433550MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 1-(1-Ethylpiperidin-3-yl)methanamine
CAS 102459-02-9
Summenformel C8H18N2
Menge 50 mg
Synonym [(1-Ethylpiperidin-3-yl)methyl]amine
InChI-Formel InChI=1S/C8H18N2/c1-2-10-5-3-4-8(6-9)7-10/h8H,2-7,9H2,1H3
SMILES CCN1CCCC(CN)C1
IUPAC-Name (1-ethylpiperidin-3-yl)methanamin
Molekulargewicht (g/mol) 142.24
Formelmasse 142.147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.