missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,6-Tetrahydrophthalsäureanhydrid-3,3,4,5,6,6-D6 (Technische Qualität), TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | 1,2,3,6-Tetrahydrophthalic Anhydride-3,3,4,5,6,6-d6 |
| CAS | 89614-23-3 |
| Summenformel | C8 2H6 H2 O3 |
| Menge | 50 mg |
| Synonym | 3a,4,7,7a-Tetrahydro-4,7-d2-1,3-isobenzofurandione-4,5,6,7-d4 (9Cl) |
| InChI-Formel | InChI=1S/C8H8O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-2,5-6H,3-4H2/i1D,2D,3D2,4D2 |
| SMILES | [2H]C1=C([2H])C([2H])([2H])C2C(C(=O)OC2=O)C1([2H])[2H] |
| IUPAC-Name | 4,4,5,6,7,7-Hexadeuterio-3a,7a-dihydro-2-benzofuran-1,3-dion |
| Molekulargewicht (g/mol) | 158.18 |
| Formelmasse | 158.085 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?