Learn More
1,8-Pyrendiamin, TRC
Specifications
Specifications
| Chemischer Name oder Material | 1,8-Pyrenediamine |
| CAS | 30269-04-6 |
| Summenformel | C16H12N2 |
| Menge | 100 mg |
| Synonym | 1,8-Diaminopyrene |
| InChI-Formel | InChI=1S/C16H12N2/c17-13-7-3-9-1-2-10-4-8-14(18)12-6-5-11(13)15(9)16(10)12/h1-8H,17-18H2 |
| SMILES | C1=CC2=C3C(=C(C=C2)N)C=CC4=C(C=CC1=C43)N |
| IUPAC-Name | Pyren-1,8-diamin |
| Molekulargewicht (g/mol) | 232.28 |
| Formelmasse | 232.1 |