Learn More
1-Benzyl-4-piperidinol-3,3,5,5-d4, TRC
Specifications
Specifications
| Chemischer Name oder Material | 1-Benzyl-4-piperidinol-3,3,5,5-d4 |
| CAS | 88227-11-6 |
| Summenformel | C12H13D4NO |
| Menge | 100 mg |
| Synonym | 1-(Phenylmethyl)-4-piperidinol-d4, 4-Hydroxy-1-benzylpiperidine-d4, 4-Hydroxy-N-benzylpiperidine-d4, N-Benzyl-4-hydroxypiperidine-d4, NSC 72991-d4, 1-(Phenylmethyl)-4-piperidin-3,3,5,5-d4-ol |
| InChI-Formel | InChI=1S/C12H17NO/c14-12-6-8-13(9-7-12)10-11-4-2-1-3-5-11/h1-5,12,14H,6-10H2/i6D2,7D2 |
| SMILES | [2H]C1([2H])CN(Cc2ccccc2)CC([2H])([2H])C1O |
| IUPAC-Name | 1-Benzyl-3,3,5,5-tetradeuteriopiperidin-4-ol |
| Molekulargewicht (g/mol) | 195.29 |
| Formelmasse | 195.1561 |