missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromotridecane-10,10,11,11,12,12,13,13,13-d9, CDN
Click to view available options
Menge:
0.1 g
0.25 g
Packungsgröße:
0.10 Gramm
0.25 Gramm
Spezifikation
Spezifikation
| Chemischer Name oder Material | 1-Bromotridecane-10,10,11,11,12,12,13,13,13-D9 |
| Summenformel | C13 D9 H18 Br |
| Menge | 0.1 g |
| Synonym | 13-Bromotridecane-1,1,1,2,2,3,3,4,4-d9, 1-Bromotridecane-10,10,11,11,12,12,13,13,13-d9, 1-Bromotridecane-10,10,11,11,12,12,13,13,13-D9, 1-Tridecyl-10,10,11,11,12,12,13,13,13-d9 bromide |
| InChI-Formel | InChI=1 S/C13H27Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14/h2-13H2,1H3/i1D3,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCCCCCCCCBr |
| IUPAC-Name | 13-bromo-1,1,1,2,2,3,3,4,4-nonadeuteriotridecane |
| Molekulargewicht (g/mol) | 272.3129 |
| Formelmasse | 271.1861 g/mol |
| Reinheit (%) | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction