missing translation for 'onlineSavingsMsg'
Learn More

1-des(piperidin-3R-amino)-1-chloro-Alogliptin, TRC

Product Code. 30353549
Change view
Click to view available options
Menge:
10 mg
100 mg
Unit Size:
100 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30353549 100 mg 100 Milligramm
30350412 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30353549 Supplier Toronto Research Chemicals Supplier No. TRCC365470100MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865758-96-9
Summenformel C13H10ClN3O2
Molekulargewicht (g/mol) 275.69
Synonym 2-[(6-Chloro-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl)methyl]-benzonitrile, Alogliptin 6-Chloro Impurity
IUPAC-Name 2-[(6-chloro-3-methyl-2,4-dioxopyrimidin-1-yl)methyl]benzonitrile
SMILES CN1C(=O)C=C(Cl)N(Cc2ccccc2C#N)C1=O

Specifications

Chemischer Name oder Material 2-[(6-Chloro-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl)methyl]-benzonitrile
Menge 100 mg
InChI-Formel InChI=1S/C13H10ClN3O2/c1-16-12(18)6-11(14)17(13(16)19)8-10-5-3-2-4-9(10)7-15/h2-6H,8H2,1H3
Formelmasse 275.0462
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.