missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthol-d8, CDN
Click to view available options
Menge:
0.25 g
0.5 g
missing translation for 'unitSize'
0.25 Gramm
0.50 Gramm
Specifications
Specifications
| Chemischer Name oder Material | 1-Naphthol-D8 |
| CAS | 207569-03-7 |
| Summenformel | C10 D8 O |
| Menge | 0.25 g |
| Synonym | 1-Naphthalen-2,3,4,5,6,7,8-d7-ol-d (9 CI), 1-Naphthol-d8, O,2,3,4,5,6,7,8-Octadeuterionaphthalen-1-ol, 1-Naphthol (OD,ring-D7), 1-Naphthol-O,2,3,4,5,6,7,8-d8, 1-Naphthol-D8 |
| InChI-Formel | InChI=1 S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D/hD |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c([2 H])c2c([2 H])c([2 H])c([2 H])c([2 H])c12 |
| IUPAC-Name | 1,2,3,4,5,6,7-heptadeuterio-8-deuteriooxynaphthalene |
| Molekulargewicht (g/mol) | 152.2192 |
| Formelmasse | 152.1077 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction