Learn More
gamma C Crystallin Antibody (OTI1E3), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP3-44646
Artikelnummer. 30306494
  • 735.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
This item is not returnable. View return policy
Description
Description
gamma C Crystallin Monoclonal antibody specifically detects gamma C Crystallin in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
gamma C Crystallin
Monoclonal
Unconjugated
Lyophilized powder ( 1X PBS, pH 7.3,)
Mouse
>95%
RUO
Primary
For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process.
Store at -20 degrees C. Avoid freeze/thaw cycles.
Lyophilized
Western Blot
OTI1E3
Western Blot 1:2000
CCL, CRYG3gamma-C-crystallin, crystallin, gamma C, crystallin, gamma-3, Gamma-C-crystallin, Gamma-crystallin 2-1, Gamma-crystallin 3, gamma-crystallin C
Full length human recombinant protein of human CRYGC (NP_066269) produced in HEK293T cell.
100 μg
Vision
1420
Human, Mouse, Rat
Antibody
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications