missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
RAB23 Monoclonal antibody specifically detects RAB23 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Spezifikation
Spezifikation
| Antigen | RAB23 |
| Anwendungen | Immunohistochemistry, Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI4C7 |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:2000, Immunohistochemistry 1:500 |
| Zusammensetzung | Lyophilized powder ( 1X PBS, pH 7.3,) |
| Gen-Alias | DKFZp781H0695, HSPC137, MGC8900, RAB family small GTP binding protein RAB 23, RAB23, member RAS oncogene family, ras-related protein Rab-23 |
| Wirtsspezies | Mouse |
| Immunogen | Human recombinant protein fragment corresponding to amino acids 1-234 of human RAB23(NP_899050) produced in E.coli. |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?