Learn More
Invitrogen™ 15-PGDH Recombinant Rabbit Monoclonal Antibody (025)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA560316
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
15-PGDH Recombinant Monoclonal Antibody for ELISA
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.
Spezifikation
| 15-PGDH | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P15428 | |
| HPGD | |
| Recombinant Human HPGD/15-PGDH protein (Met1-Gln266). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 025 | |
| Unconjugated | |
| HPGD | |
| 15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NAD(+)]; 15-PGDH; AV026552; HPGD; hydroxyprostaglandin dehydrogenase 15 (NAD); hydroxyprostaglandin dehydrogenase 15-(NAD); NAD+-dependent 15-hydroxyprostaglandin dehydrogenase; NAD-dependent 15-hydroxyprostaglandin dehydrogenase; PGDH; PGDH1; PHOAR1; Prostaglandin dehydrogenase 1; SDR36C1; Short chain dehydrogenase/reductase family 36C member 1; short chain dehydrogenase/reductase family 36C, member 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3248 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |