Learn More
Testo™ TEMPERATURE DATALOGGER 2 K CHANNEL CHANNEL
Artikelnummer. 11351374
Ansicht ändern
Click to view available options
Kanäle:
2 Extern
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Kanäle Packungsgröße
11351374 2 Extern 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11351374 Lieferant Testo™ Lieferanten-Nr. 0572 1753
  • ONLINE EXCLUSIVE PRICE
    408.60 €
    243.00 € / 1 Stück
    Sparen 165.60 €
    40% Rabatt
Verfügbar ab: 19-01-2027