missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6,10,15,19,23-Hexamethyltetracosane-d62, CDN
Click to view available options
Menge:
0.5 g
1 g
Packungsgröße:
0.50 Gramm
1 Gramm
Spezifikation
Spezifikation
| Chemischer Name oder Material | 2,6,10,15,19,23-Hexamethyltetracosane-d62 |
| CAS | 16514-83-3 |
| Summenformel | C30 D62 |
| Menge | 1 g |
| Synonym | Tetracosane-1,1,1,2,3,3,4,4,5,5,6,7,7,8,8,9,9,10,11,11,12,12,13,13,14,14,15,16,16,17,17,18,18,19,20,20,21,21,22,22,23,24,24,24-d44, 2,6,10,15,19,23-hexa(methyl-d3)- (8 CI, 9 CI, ACI), 2,6,10,15,19,23-Hexa(methyl-d3)tetracosane-1,1,1,2,3,3,4,4,5,5,6,7,7,8,8,9,9,10,11,11,12,12,13,13,14,14,15,16,16,17,17,18,18,19,20,20,21,21,22,22,23,24,24,24-d44 (ACI), Perdeuteriosqualane, Squalane (C30D62), 2,6,10,15,19,23-Hexamethyltetracosane-d62, Squalane-D62 |
| InChI-Formel | InChI=1 S/C30H62/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h25-30 H,9-24H2,1-8H3/i1D3,2D3,3D3,4D3,5D3,6D3,7D3,8D3,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25 D,26 D,27 D,28 D,29 D,30 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC-Name | 1,1,1,2,3,3,4,4,5,5,6,7,7,8,8,9,9,10,11,11,12,12,13,13,14,14,15,16,16,17,17,18,18,19,20,20,21,21,22,22,23,24,24,24-tetratetracontadeuterio-2,6,10,15,19,23-hexakis(trideuteriomethyl)tetracosane |
| Molekulargewicht (g/mol) | 485.1953 |
| Formelmasse | 484.8743 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction