missing translation for 'onlineSavingsMsg'
Learn More

rac Zearalenone-d6, TRC

Artikelnummer. 30454076
Click to view available options
Menge:
1 mg
Unit Size:
1 Milligramm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30454076

Brand: Toronto Research Chemicals TRCZ2705021MG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material rac Zearalenone-d6
CAS 1185236-04-7
Summenformel C18H16D6O5
Menge 1 mg
Synonym 10,10,11-Trideuterio-15,17-dihydroxy-11-(trideuteriomethyl)-12-oxabicyclo[12.4.0]octadeca-1(18),2,14,16-tetraene-7,13-dione, rac-Zearalenone-d6, rac-Zearalenone-D6 (10-hydroxy-6-oxo-1-undecenyl-9,9,10,11,11,11-D6), (3RS,11EZ)-3,4,5,6,9,10-Hexahydro-14,16-dihydroxy-3-(methyl-d3)-1H-2-benzoxacyclotetradecin-1,7(8H)-dione-3,4,4-d3, 6-(10-Hydroxy-6-oxo-1-undecenyl-9,9,10,11,11,11-d6)-β-resorcylic acid lactone
InChI-Formel InChI=1S/C18H22O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,20-21H,2,4-6,8-9H2,1H3/i1D3,6D2,12D
SMILES [2H]C([2H])([2H])C1([2H])OC(=O)c2c(O)cc(O)cc2C=CCCCC(=O)CCC1([2H])[2H]
IUPAC-Name 10,10,11-trideuterio-15,17-dihydroxy-11-(trideuteriomethyl)-12-oxabicyclo[12.4.0]octadeca-1(14),2,15,17-tetraene-7,13-dione
Molekulargewicht (g/mol) 324.4
Formelmasse 324.1844
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.