missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)-1-propanol, TRC

Product Code. 30476201
Change view
Click to view available options
Menge:
25 mg
Unit Size:
25 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30476201 25 mg 25 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476201 Supplier Toronto Research Chemicals Supplier No. TRCN37315525MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 2-(1-Naphthyl)-1-propanol
CAS 101349-60-4
Summenformel C13H14O
Menge 25 mg
Synonym beta-Methyl-1-naphthaleneethanol
InChI-Formel InChI=1S/C13H14O/c1-10(9-14)12-8-4-6-11-5-2-3-7-13(11)12/h2-8,10,14H,9H2,1H3
SMILES CC(C(=O)O)c1cccc2ccccc12
Molekulargewicht (g/mol) 186.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.