Learn More
2,2'-Methylenbis(4-bromphenol), TRC
Specifications
Specifications
| Chemischer Name oder Material | 2,2'-Methylenebis(4-bromophenol) |
| CAS | 78563-03-8 |
| Summenformel | C24 H34 O4 |
| Menge | 5 g |
| Synonym | Bis(2-hydroxy-5-bromophenyl)methane |
| InChI-Formel | InChI=1S/C24H34O4/c1-24-11-10-17-16-7-6-15(26-2)13-19(16)21(25)14-18(17)20(24)8-9-22(24)28-23-5-3-4-12-27-23/h6-7,13,17-18,20-23,25H,3-5,8-12,14H2,1-2H3/t17-,18+,20+,21?,22-,23?,24-/m0/s1 |
| SMILES | COc1ccc2[C@@H]3CC[C@]4(C)[C@H](CC[C@@H]4[C@@H]3CC(O)c2c1)OC5CCCCO5 |
| IUPAC-Name | (8R,9R,13S,14R,17S)-3-Methoxy-13-methyl-17-(oxan-2-yloxy)-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-6-ol |
| Molekulargewicht (g/mol) | 386.524 |
| Formelmasse | 386.246 |