missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Tribromiodbenzol, TRC

Artikelnummer. 30450602
Ansicht ändern
Click to view available options
Menge:
100 mg
250 mg
Packungsgröße:
100 Milligramm
250 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30450602 100 mg 100 Milligramm
30450827 250 mg 250 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30450602 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCT366490100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 2,4,5-Tribromoiodobenzene
CAS 366496-32-4
Summenformel C6H2Br3I
Menge 100 mg
Synonym 1,2,4-Tribromo-5-iodobenzene
InChI-Formel InChI=1S/C6H2Br3I/c7-3-1-5(9)6(10)2-4(3)8/h1-2H
SMILES Brc1cc(Br)c(I)cc1Br
IUPAC-Name 1,2,4-Tribrom-5-iodbenzol
Molekulargewicht (g/mol) 440.7
Formelmasse 437.6751
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.