missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-dichloropyridine-3-boronic acid, 95%, Maybridge
Click to view available options
Menge:
1 g
Verpackung:
Amber Glass Bottle
Spezifikation
Chemischer Name oder Material | 2, 6-Dichloropyridine-3-boronic acid |
CAS | 148493-34-9 |
CAS Min. % | 95.0 |
Prozentgehaltsbereich | 95% |
Verpackung | Amber Glass Bottle |
Summenformel | C5H4BCl2NO2 |
MDL-Nummer | MFCD03094989 |
Menge | 1 g |
Synonym | 2,6-dichloropyridine-3-boronic acid, 2,6-dichloropyridin-3-yl boronic acid, 2,6-dichloro-3-pyridineboronic acid, 3-borono-2,6-dichloropyridine, 2,6-dichloropyridin-3-yl-3-boronic acid, 2,6-dichloro-3-pyridinyl-boronic acid, boronic acid, 2,6-dichloro-3-pyridinyl, 2,6-dichloropyridine-3-boronicacid, pubchem5039, acmc-209d0p |
InChI-Schlüssel | XBBLBQZAVMHEER-UHFFFAOYSA-N |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'